Would you like to log in again?
Safety Shower Stations, WaterSaver Faucet
Supplier: WaterSaver Faucet
SSC902 SS902 SSCBF909 SSC909 CTSSBF919 CTSS912 CTSS919
56611-008EA 1511.39 USD InStock
56611-008 VFCTSS919 VFCTSSBF919 59380-036 VFCTSS912 59380-038 59380-034
Safety Shower Stations, WaterSaver Faucet
First Aid Supplies Showers
Designed to provide complete personal protection, these corrosion-resistant safety stations feature a single water inlet that supplies both the shower and eye wash.
  • Supplied with rigid stainless steel pull rods
  • Self-regulating flow controls are positioned inside each spray head
  • Stations are provided with an ANSI-compliant sign
  • 2 versions available: Orange ABS plastic shower head and galvanized steel pipes and fittings with orange PE covers or cast brass showerhead and white epoxy/polyester hybrid powder coated brass pipe and fittings
The high volume deluge shower head has a minimum 75.7 L/min. (20 gpm) flow and 50.8 cm (20") spray pattern.
Certifications: Meets ANSI Z358.1 standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty