Would you like to log in again?
VWR® Contour™ Metal Wall Angles
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR® Contour™
VWL-2030-00 VFL-2030-00
VWL-2030-00EA 179.48 USD InStock
VWL-2030-00 VFL-2030-00
VWR® Contour™ Metal Wall Angles
Furniture Casework
Made of 16-gauge metal, wall angles attach to the wall to support the rear overhang of worktops and wall table service ledges.
Ordering information: Metal wall angle VFL-2030-00 is shipped in two 122 cm (48") lengths.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty