Would you like to log in again?
Panel-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
L511WS L512VB-WS L511VB-WS L512WS
VFL512VB-WSEA 569.56 USD InStock
VFL512VB-WS VFT-0354-6V VFL512WS VFT-0354-06
Panel-Mounted Gooseneck Mixing Faucets, with Four-Arm Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Panel-mounted mixing faucets are made of chrome-plated cast brass and provided with a serrated hose end, four-arm handles, and 1 cm (3/8") IPS inlet shanks, locknuts, and washers
Faucets are available with 15.2 cm (6") spread gooseneck, with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty