Would you like to log in again?
Deck-Mounted Gooseneck Mixing Faucets with Vacuum Breaker
Supplier: WaterSaver Faucet
L412-12VB L411-8VB L414-12VB L412-8VB L411VB L412VB
VFL412-8VBEA 405.46 USD InStock
VFL412-8VB 48888-044 VFL412VB VFL414-12VB VFT-0340-OV 59382-044
Deck-Mounted Gooseneck Mixing Faucets with Vacuum Breaker
Faucets and Sinks Faucets
Deck-mounted mixing faucets are made of chrome-plated forged brass and provided with a serrated hose end, four-arm handles, and 1 cm (³/₈") diameter copper tubes on inlets.
  • Faucet valves are self-contained renewable units with replaceable stainless steel seats
Faucets are available with 15.2 cm (6"), 20.3 cm (8"), or 30.5 cm (12") spread gooseneck, all with a vacuum breaker.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Accessories information: Chrome-plated brass faucet accessories include an aerator and an aspirator faucet outlet.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty