Would you like to log in again?
Panel-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Supplier: WaterSaver Faucet
CT1714WS CT1714VB-WSA
VFT-1714-00EA 383.37 USD InStock
VFT-1714-00 VFCT1714VB-WSA
Panel-Mounted Gooseneck Mixing Faucets, with Nylon Hooded Handles, WaterSaver Faucet
Faucets and Sinks Faucets
Panel-mounted mixing faucets are made of white epoxy/polyester hybrid powder-coated cast brass
They are provided with a serrated hose end, black nylon hooded handles, and 1 cm (3/8") mounting shanks, locknuts, and washers. Horizontal inlets are on 15.2 cm (6") centers and gooseneck spread is 15.2 cm (6"). Available with or without a vacuum breaker. Faucet valves are self-contained renewable units with replaceable stainless steel seats.
Certifications: CSA certified. Comply with ANSI/ASME A112.18.1M.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty