Would you like to log in again?
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Supplier: BD Medical
BD Vacutainer®
368650 368607 368608 368651 368650 368608 368651 368607
80089-444CS 104.88 USD InStock
CABD368607L 80089-444 CABD368650 VT-368607 CABD368608L CABD368651 VT-368608 80089-442
BD Vacutainer® Eclipse™ Blood Collection Needles, BD Diagnostics
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These sterile, safety-engineered, multi-sample blood collection needles offer a simple, effective way to collect blood while reducing the possibility of needlestick injuries
They feature a patented safety shield that allows for one-handed activation to cover the needle completely upon withdrawal from the vein, and confirm proper activation with an audible click. The safety shield is already aligned with the bevel of the needle, so no extra assembly or adjustment is required by the user. The intuitive design of the advanced protection technology means there is no need to change the standard, one-handed venipuncture technique when using these needles. Needles with pre-attached holders help facilities remain single-use holder compliant.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty