Would you like to log in again?
VWR® Atlas Metal Shelving, Single Lip
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
VWR1L1872 VWR1L1542 VWR1L1848 VWR1L1566 VWR1L1572 VWR1L2460-W VWR1L1554-W VWR1L1260 VWR1L1248 VWR1L1260-W VWR1L1554 VWR1L2454-W VWR1L1242 VWR1L1254-W VWR1L1548 VWR1L1542-W VWR1L2454 VWR1L1254 VWR1L2448 VWR1L1860-W VWR1L1236-W VWR1L1236 VWR1L2442 VWR1L1266 VWR1L2460 VWR1L2436 VWR1L2436-W VWR1L1860 VWR1L1836 VWR1L1272 VWR1L1872-W VWR1L1836-W VWR1L1854-W VWR1L2448-W VWR1L2472-W VWR1L1272-W VWR1L1866 VWR1L1536-W VWR1L1560 VWR1L1242-W VWR1L1248-W VWR1L1572-W VWR1L1848-W VWR1L1548-W VWR1L1560-W VWR1L1536 VWR1L1842 VWR1L1854 VWR1L2466 VWR1L2442-W VWR1L2472 VWR1L1266-W
VWR1L2460-WEA 520.18 USD InStock
VWR1L2460-W VWR1L1248-W VWR1L2466 VWR1L1254 VWR1L1242-W VWR1L1572 VWR1L2460 VWR1L1854-W VWR1L1272-W VWR1L1854 VWR1L1536 VWR1L1560-W VWR1L2472-W VWR1L1860-W VWR1L1236-W VWR1L2454 VWR1L1560 VWR1L1242 VWR1L1554-W VWR1L1848 VWR1L1836-W VWR1L1248 VWR1L2436-W VWR1L1566 VWR1L1842 VWR1L1872 VWR1L1266-W VWR1L1554 VWR1L2442 VWR1L1872-W VWR1L1272 VWR1L1260-W VWR1L1542-W VWR1L1836 VWR1L1848-W VWR1L2448-W VWR1L2448 VWR1L1236 VWR1L1548-W VWR1L2442-W VWR1L1260 VWR1L2454-W VWR1L1542 VWR1L1266 VWR1L1254-W VWR1L1860 VWR1L2472 VWR1L1548 VWR1L1866 VWR1L1536-W VWR1L2436 VWR1L1572-W
VWR® Atlas Metal Shelving, Single Lip
Furniture Shelves
Designed to be infinitely adjustable along Atlas uprights.
  • Standard shelves are powder-coated with high-quality, chemical-resistant epoxy powder
  • Constructed of 16-gauge steel
Lipped shelves can be used with the lip in the front or back and can be switched in the field. Light neutral shelves are 3 cm (13/16") thick with 1.1 cm (7/16") high lips.
Ordering information: Shelf brackets are available separately; see VWRBHC12 series.
Order Now
ORDER
ACCESSORIES
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty