Would you like to log in again?
VWR® Single-Lipped Atlas Shelves
Supplier: VWR
VWR®
VWR1L1530-W VWR1L2430 VWR1L1530 VWR1L1230-W VWR1L1830 VWR1L1230 VWR1L1830-W
VWR1L1830EA 282.34 USD InStock
VWR1L1830 VWR1L1830-W VWR1L1230-W VWR1L1530 VWR1L2430 VWR1L1530-W VWR1L1230
VWR® Single-Lipped Atlas Shelves
Furniture Shelves
These standard shelves are powder-coated with high-quality, chemical-resistant epoxy powder and constructed of 16-gauge steel.
  • Chemical-resistant epoxy powder coating
  • Steel construction
  • Single-lipped design
Designed to be infinitely adjustable along Atlas uprights, these lipped shelves can be used with the lip in the front or back and can be switched in the field. Shelf only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty