Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Atlas Metal Shelving, Double Lip

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
VWR2L2460 VWR2L1236 VWR2L2442 VWR2L1266 VWR2L1260-W VWR2L1842-W VWR2L1536 VWR2L2448 VWR2L1560 VWR2L1248-W VWR2L1836 VWR2L1866 VWR2L1566 VWR2L2442-W VWR2L1266-W VWR2L1254 VWR2L1242 VWR2L2448-W VWR2L1860 VWR2L2454 VWR2L1860-W VWR2L1848 VWR2L1248 VWR2L1272 VWR2L1554 VWR2L1548 VWR2L2436 VWR2L1548-W VWR2L1848-W VWR2L1542 VWR2L2460-W VWR2L1272-W VWR2L2454-W VWR2L1854 VWR2L2472-W VWR2L1242-W VWR2L1842 VWR2L2472 VWR2L2466 VWR2L1536-W VWR2L1260
VWR2L2460-WEA 558.56 USD InStock
VWR2L2460-W VWR2L1248-W VWR2L2442-W VWR2L1536 VWR2L2448 VWR2L2448-W VWR2L1854 VWR2L1242-W VWR2L1836 VWR2L2442 VWR2L1272-W VWR2L1254 VWR2L1554 VWR2L1848-W VWR2L1236 VWR2L2466 VWR2L1548-W VWR2L2460 VWR2L1272 VWR2L2472-W VWR2L1860-W VWR2L2454-W VWR2L1866 VWR2L1548 VWR2L1248 VWR2L1842 VWR2L2436 VWR2L1848 VWR2L1842-W VWR2L1260-W VWR2L1242 VWR2L1560 VWR2L2472 VWR2L1566 VWR2L1536-W VWR2L1266 VWR2L1266-W VWR2L1860 VWR2L2454 VWR2L1542 VWR2L1260
VWR® Atlas Metal Shelving, Double Lip
Furniture Shelves
Designed to be infinitely adjustable along Atlas uprights.

  • Shelves are powder-coated with high-quality, chemical-resistant epoxy powder
  • Constructed of 16-gauge steel

Light neutral shelves are 3 cm (13/16") thick with 1.1 cm (7/16") high lips.

Ordering information: Shelf brackets are available separately; see VWRBHC12 series.

Order Now
 

ORDER

ACCESSORIES

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty