Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Atlas Overhead Storage Cabinets with Sliding Doors

Supplier: VWR

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

VWR®
VWR407521A VWR407520A-W VWR407525M VWR407525G VWR407522M-W VWR407526A VWR407520M VWR407526G VWR407520G VWR407526M VWR407521G VWR407522G VWR407520M-W VWR407522A VWR407525A VWR407523M VWR407522M VWR407521M VWR407523A VWR407523G VWR407520A VWR407524A VWR407524G VWR407524M VWR407521M-W VWR407520G-W
VWR407524GEA 1670 USD InStock
VWR407524G VWR407526G VWR407520A VWR407520G VWR407526M VWR407522G VWR407521M-W VWR407526A VWR407522A VWR407524A VWR407521M VWR407523M VWR407523G VWR407520G-W VWR407525G VWR407520M-W VWR407525M VWR407522M-W VWR407521G VWR407520A-W VWR407525A VWR407521A VWR407523A VWR407522M VWR407524M VWR407520M
VWR® Atlas Overhead Storage Cabinets with Sliding Doors
Furniture Storage Cabinets
Steel cabinets are used in place of open shelving when enclosed storage and/or resistance to dust is required.

  • Sliding doors
  • Color: Light neutral

For double-sided storage without pass-through, two cabinets of the same size may be mounted back to back.

For use with VWR® Atlas mobile carts/mobile laboratory benches.

Ordering information: Supplied with mounting hardware. Hardware for back-to-back installation not included.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty