Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

VWR® Atlas Pass-Through Overhead Storage Cabinets with Hinged Doors

Supplier: VWR
VWR®
VWR407534HG VWR407535HM VWR407531HM VWR407533HG VWR407532HM VWR407534HM VWR407532HG VWR407531HG VWR407536HM VWR407533HM VWR407530HG VWR407530HM
VWR407533HGEA 7022.67 USD InStock
VWR407533HG VWR407534HG VWR407531HG VWR407532HG VWR407531HM VWR407532HM VWR407530HM VWR407535HM VWR407536HM VWR407533HM VWR407534HM VWR407530HG
VWR® Atlas Pass-Through Overhead Storage Cabinets with Hinged Doors
Furniture Storage Cabinets
Steel cabinets are used in place of open shelving when enclosed storage and/or resistance to dust is required.

  • Steel fabrication
  • Hinged doors
  • Color: Light neutral

For use with VWR® Atlas mobile carts/mobile laboratory benches.

Ordering information: Supplied with two adjustable shelves and mounting hardware.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty