Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium dihydrogen phosphate monohydrate USP endotoxin tested

Supplier: Avantor Performance Materials
Synonyms: Sodium dihydrogen orthophosphate 1-hydrateSodium dihydrogen orthophosphate monohydrateSodium dihydrogen phosphate 1-hydrateSodium dihydrogenorthophosphate monohydrateSodium orthophosphate monobasic monohydrateSodium phosphate monobasic monohydrate

VWRVVWRB0823-12KG VWRVVWRB0823-1KG
VWRB0823-1KGEA 478.59 USD InStock
VWRB0823-1KG VWRB0823-12KG
Sodium dihydrogen phosphate monohydrate USP endotoxin tested
Sodium dihydrogen phosphate monohydrate
Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.


Formula: NaH₂PO₄·H₂O
MW: 137.99 g/mol
Melting Pt: 100 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149208
CAS Number: 10049-21-5
EINECS: 231-449-2
Merck Index: 12,08806
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/57/16640557.jpg
Order Now
Specification Test Results
Aluminum, Calcium & Related Elements PASS / Fail
Arsenic ≤ 8 ppm
Chloride ≤ 0.014%
Endotoxin 2.5000 EU/g
Heavy Metals ≤ 20 ppm
Identification (A) PASS / Fail
Identification (B) PASS / Fail
Insoluble Substances ≤ 0.2 %
pH (1.0g/20mL Water) @25C 4.1 - 4.5
Purity (Anhydrous Basis) 98.0 - 103.0%
Sulfate ≤ 0.15%
Water Determination 10.0 - 15.0%
Additional information, country of origin Germany
Additional information Material is of non-animal, non-human origin
Additional information, storage conditions 15ºC to 30ºC

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty