Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Guanidinium hydrochloride, Bioreagent

Supplier: Avantor Performance Materials
Warning
Synonyms: Guanidinium chlorideGuanidine hydrochlorideGuanidine chlorideGuanadine hydrochloride

VWRVVWRB287-12KG VWRVVWRB287-5KG VWRVVWRB287-50KG
VWRB287-5KGEA 2189.43 USD InStock
VWRB287-5KG VWRB287-50KG VWRB287-12KG
Guanidinium hydrochloride, Bioreagent
Guanidinium hydrochloride
Material is of non-animal, non-human origin.

  • Bioprocessing
  • Bioreagent


Formula: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Melting Pt: 181…187 °C
Density: 1.354 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013026
CAS Number: 50-01-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/24/2297424.jpg
Order Now
Specification Test Results
Abs. 230nm (6M, Water) ≤ 0.2
Abs. 260nm (6M, Water) ≤ 0.03
Abs. 275nm (6M, Water) ≤ 0.03
Arsenic ≤ 0.0500 ppm
Assay 99.5-101.0%
Copper ≤ 0.200 ppm
Cyanide ≤ 0.1 ppm
Dnase None
Iron ≤ 2.0 ppm
Lead ≤ 0.500 ppm
Loss on Drying ≤ 0.5%
Nitrate ≤ 0.005%
Protease None
Residue on Ignition ≤ 0.05%
Solubility (6M, Water) Pass
Sulfate ≤ 0.005%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty