Synonyms:
Carbamide Carbonyldiamide
VWRVVWRB568-12KG
VWRVVWRB568-500G
VWRVVWRB568-50KG
VWRB568-12KGEA
1089.01
USD
InStock
VWRB568-12KG
VWRB568-500G
VWRB568-50KG
Urea 98.0-102.0% USP, Ph. Eur., BP, JP endotoxin tested
Urea
Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available.