Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate, anhydrous ≥99.0% USP, Ph. Eur. endotoxin tested

Supplier: Avantor Performance Materials
Synonyms: Bisodium sulfateDisodium sulfateSulfuric acid disodium salt

VWRVVWRB836-500G VWRVVWRB836-12KG
VWRB836-12KGEA 933.49 USD InStock
VWRB836-12KG VWRB836-500G
Sodium sulfate, anhydrous ≥99.0% USP, Ph. Eur. endotoxin tested
Sodium sulphate
Suitable for bioprocessing. BSE/TSE free. Endotoxin tested. Custom packaging and testing available. Change management and notification available. Samples available

Material is of non-animal, non-human origin


Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 888 °C
Density: 2.7 g/cm³ (20 °C)
MDL Number: MFCD00003504
CAS Number: 7757-82-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg
Order Now
Specification Test Results
Acidity or Alkalinity (USP) PASS
Assay (USP) ≥99.0%
Endotoxins (USP) ≤2.5 EU/g
Chloride (USP) 221 ≤0.02%
Heavy Metals (USP) 231 ≤0.001%
Identification (USP) PASS
Loss on Drying (USP) 731 ≤0.5%
Acidity or Alkalinity (EP) PASS
Appearance of Solution (EP) PASS
Assay (EP) 98.5 – 101.0%
Calcium (EP) (243) ≤ 450ppm
Chlorides (EP) (244) ≤ 450ppm
Heavy Metals (EP) (248) ≤ 45ppm
Identification A (EP) PASS
Identification B (EP) PASS
Identification C (EP) PASS
Iron (EP) (249) ≤ 90ppm
Loss on Drying (EP) (2232) ≤ 0.5%
Magnesium (EP) ≤ 200ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty