Would you like to log in again?
VWR® Atlas Pegboards
Supplier: VWR
VWR®
VWRBCP3048 VWRBCP3648 VWRECP2932 VWRBCP3636 VWRBCPMB VWRBCP3036 VWRECP2920
VWRBCPMBPK 682 USD InStock
VWRBCPMB VWRBCP3048 VWRBCP3036 VWRBCP3648 VWRECP2932 VWRBCP3636 VWRECP2920
VWR® Atlas Pegboards
Furniture Shelves
Post-mounted epoxy pegboards are 2.5 cm (1") thick and sized to mount between standard 91.4 cm (36") and 121.9 cm (48") center-to center dimensions of Atlas uprights.
  • Adjustable pegboards
  • Color: Black
Pegboards are adjustable in height, allowing counters to remain clear of clutter. Pegboards feature black pegs, a stainless steel trough, and a finished back.
Ordering information: Mounting brackets VWRBCPMB are required for installation and must be purchased separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty