Would you like to log in again?
VWR® Atlas Shelf Brackets
Supplier: VWR
VWR®
VWRBLC18 VWRBLC24 VWRBL15 VWRBLC15-W VWRBHC24 VWRBH18-W VWRBLC18-W VWRBU12 VWRBL15-W VWRBU24-W VWRBH24 VWRBLC36 VWRBU36 VWRBH24-W VWRBHC12 VWRBLC24-W VWRBHC15-W VWRBHC18-W VWRBH15-W VWRBHC15 VWRBLC15 VWRBH12 VWRBL12-W VWRBU18-W VWRBH15 VWRBU18 VWRBHC12-W VWRBH18 VWRBU36-W VWRBLC12 VWRBHC18 VWRBL18 VWRBL24 VWRBL24-W VWRBU24 VWRBH12-W VWRBLC12-W VWRBL12 VWRBU12-W
VWRBL15-WEA 199.56 USD InStock
VWRBL15-W VWRBHC15 VWRBHC18 VWRBU12-W VWRBH24 VWRBL24 VWRBL12-W VWRBHC18-W VWRBU18-W VWRBL18 VWRBH12-W VWRBU12 VWRBLC24-W VWRBH15-W VWRBU36 VWRBHC15-W VWRBH18-W VWRBLC18 VWRBHC12 VWRBU36-W VWRBLC15 VWRBHC12-W VWRBU18 VWRBLC36 VWRBLC12 VWRBH12 VWRBH15 VWRBL12 VWRBH18 VWRBL15 VWRBL24-W VWRBH24-W VWRBLC12-W VWRBLC15-W VWRBU24 VWRBU24-W VWRBLC18-W VWRBHC24 VWRBLC24
VWR® Atlas Shelf Brackets
Furniture Shelves
Designed to be infinitely adjustable along vertical Atlas uprights.
  • Brackets are powder-coated with high-quality chemical-resistant epoxy powder
  • Constructed of 12-gauge steel
Shelf brackets provide a sturdy base of support for shelving units and are adjustable along vertical Atlas uprights.
Ordering information: Brackets are sold in sets of two. Low-profile shelf brackets and Kableway shelf brackets are designed for under mount only. For Atlas uprights, see VWRWMUP24, VWRC24, and VWRSU24 series. For Atlas shelving, see VWRNL1236, VWR1L1236, and VWR2L1236 series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty