Would you like to log in again?
VWR® Atlas Single Uprights
Supplier: VWR
VWR®
VWRC96-W VWRC72 VWRC72-W VWRC36-W VWRC66 VWRC36 VWRC48-W VWRC24 VWRC84 VWRC60 VWRC30-W VWRC66-W VWRC48 VWRC96 VWRC54 VWRC84-W VWRC30 VWRC24-W VWRC54-W VWRC60-W
VWRC54EA 805 USD InStock
VWRC54 VWRC30-W VWRC96-W VWRC84-W VWRC54-W VWRC66 VWRC36 VWRC24 VWRC66-W VWRC24-W VWRC36-W VWRC48 VWRC60-W VWRC72-W VWRC48-W VWRC72 VWRC60 VWRC96 VWRC30 VWRC84
VWR® Atlas Single Uprights
Furniture Shelves
Counter-mounted Atlas upright supports are designed to bolt directly to a new or existing countertop to provide infinitely adjustable shelving above the bench.
  • Light neutral supports with black metal inserts
These Atlas single uprights are built of durable materials.
Ordering information: Supplied with mounting hardware. Uprights 152.4 cm (60") and higher are supplied with a ceiling trim ring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty