Would you like to log in again?
VWR® Cryo Coder Cap Inserts
Supplier: VWR
VWR®
52311A1 5232601 5231151 5232551 5232571 5231121 5231111 5231131 5232581 5232591 5232561 5231141
VWRI210-000037CS 35.52 USD InStock
VWRI210-000037 VWRI210-000038 VWRI210-000039 VWRI210-000033 VWRI210-000034 VWRI210-000035 VWRI210-000036 VWRI210-000040 VWRI210-000041 VWRI210-000042 VWRI210-000031 VWRI210-000032
VWR® Cryo Coder Cap Inserts
Cap Inserts
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple color options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty