Would you like to log in again?
VWR® Atlas Metal Shelving
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
VWRNL1530 VWRNL1554 VWRNL1260 VWRNL1230 VWRNL1272 VWRNL1548 VWRNL1554-W VWRNL1566 VWRNL2448 VWRNL1530-W VWRNL1542 VWRNL1254-W VWRNL1272-W VWRNL1260-W VWRNL1266-W VWRNL2442 VWRNL1854 VWRNL1830 VWRNL1248-W VWRNL1848-W VWRNL1242-W VWRNL1236 VWRNL1254 VWRNL1536-W VWRNL1560-W VWRNL1536 VWRNL1560 VWRNL2448-W VWRNL1842 VWRNL1248 VWRNL1242 VWRNL1236-W VWRNL1860 VWRNL1860-W VWRNL1548-W VWRNL1854-W VWRNL1866 VWRNL2454 VWRNL1266 VWRNL2430 VWRNL1848 VWRNL1830-W VWRNL2436 VWRNL1836 VWRNL2460 VWRNL1572
VWRNL2460EA 528 USD InStock
VWRNL2460 VWRNL1272 VWRNL1830-W VWRNL1536-W VWRNL1854-W VWRNL2442 VWRNL1572 VWRNL1254 VWRNL2448-W VWRNL1230 VWRNL1554 VWRNL1830 VWRNL1848-W VWRNL1236 VWRNL1530 VWRNL1536 VWRNL2448 VWRNL1272-W VWRNL1854 VWRNL1254-W VWRNL1836 VWRNL1560-W VWRNL1236-W VWRNL1260 VWRNL1548-W VWRNL1242 VWRNL1860-W VWRNL2430 VWRNL1560 VWRNL1530-W VWRNL1566 VWRNL1860 VWRNL1266 VWRNL1542 VWRNL2454 VWRNL1866 VWRNL1548 VWRNL1260-W VWRNL1842 VWRNL1248 VWRNL1554-W VWRNL2436 VWRNL1242-W VWRNL1848 VWRNL1266-W VWRNL1248-W
VWR® Atlas Metal Shelving
Furniture Shelves
Designed to be infinitely adjustable along Atlas uprights.
  • Constructed of 16-gauge steel
  • Shelves are powder-coated with high-quality, chemical-resistant epoxy powder
Ordering information: Shelf brackets are available separately; see VWRBHC12 series.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty