Would you like to log in again?
VWR® Atlas Center-Mounted Umbrella Shelving
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
VWR®
VWRNLU1848-W VWRNLU2436-W VWRNLU3660-W VWRNLU1242 VWRNLU3642 VWRNLU1866-W VWRNLU2436 VWRNLU2442 VWRNLU3660 VWRNLU1848 VWRNLU2454 VWRNLU1272-W VWRNLU1860 VWRNLU1254-W VWRNLU2448-W VWRNLU3648-W VWRNLU1854 VWRNLU1254 VWRNLU3672-W VWRNLU3648 VWRNLU2472-W VWRNLU2466-W VWRNLU1836-W VWRNLU1236 VWRNLU1842 VWRNLU1248 VWRNLU2442-W VWRNLU2460 VWRNLU1266 VWRNLU1866 VWRNLU2460-W VWRNLU1260-W VWRNLU1854-W VWRNLU3666 VWRNLU3654 VWRNLU1272 VWRNLU1260 VWRNLU3636 VWRNLU3642-W VWRNLU2472 VWRNLU2454-W VWRNLU2448 VWRNLU2466 VWRNLU1872 VWRNLU3672 VWRNLU1248-W VWRNLU1836 VWRNLU1236-W
VWRNLU1836-WEA 434 USD InStock
VWRNLU1836-W VWRNLU2436 VWRNLU1842 VWRNLU1248 VWRNLU3666 VWRNLU1866 VWRNLU1272-W VWRNLU3660-W VWRNLU3648 VWRNLU2442-W VWRNLU1260 VWRNLU3660 VWRNLU1854-W VWRNLU2472 VWRNLU2466-W VWRNLU1242 VWRNLU3642 VWRNLU2454 VWRNLU1860 VWRNLU2448-W VWRNLU1266 VWRNLU1236-W VWRNLU2460-W VWRNLU1848 VWRNLU1254-W VWRNLU3642-W VWRNLU3648-W VWRNLU2466 VWRNLU1872 VWRNLU3654 VWRNLU1236 VWRNLU1260-W VWRNLU1854 VWRNLU3672-W VWRNLU2448 VWRNLU3636 VWRNLU1848-W VWRNLU2454-W VWRNLU1866-W VWRNLU2460 VWRNLU1272 VWRNLU2436-W VWRNLU3672 VWRNLU2442 VWRNLU1254 VWRNLU1248-W VWRNLU2472-W VWRNLU1836
VWR® Atlas Center-Mounted Umbrella Shelving
Furniture Shelves
Umbrella shelves are mounted at the top of uprights and replace a cross brace when used.
  • 16-gauge steel construction
  • Coated with chemical-resistant epoxy powder
Shelves are powder-coated with high-quality chemical-resistant epoxy powder and constructed of 16-gauge steel.
Ordering information: Shelf brackets are available separately; see VWRBHC12 series.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty