Would you like to log in again?
xTIP® Pipette Tips for Rainin LTS Style Pipettors, Biotix
Supplier: Biotix
R-0200-9FC R-0020-9NC R-0250-9NC R-0250-9SC R-0020-9FC R-0020-9SC R-1000-9NC R-1000-9FC R-1000-9SC
75848-654CS 231.44 USD InStock
75848-654 10834-078 10834-076 10834-086 10834-074 75848-656 10834-084 10834-072 75848-658
xTIP® Pipette Tips for Rainin LTS Style Pipettors, Biotix
xTIP® Pipette Tips are especially designed for Rainin Pipets, and feature innovations beyond ergonomics
  • Experience the perfect fit with every tip using the patented FlexFit® technology, which features a flexible proximal end to ensure confidence in a secure seal.
  • Clean delivery with X-Resin® and Blade® technologies:
  • Most low retention pipette tips are unevenly coated. X-Resin® is a sleek resin that needs no coating, resulting in uniform composition throughout xTIP® tips from proximal end to distal end. X-Resin® provides ultra-low retentive qualities, with every lot, from the first tip to the last.
  • Blade® technology eliminates hanging droplet formation, ensuring sample precision with each and every delivery by reducing surface tension at the distal end of the tip.
Certified free of RNase, DNase, Pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for accuracy and precision, ensuring superior quality and compatibility. All raw materials are controlled.
FlexFit®, X-Resin®, and Blade® technologies developed by Biotix are found exclusively in Biotix and xTIP® brand pipette tips. Biotix technologies minimize inaccuracies associated with the number one issue in pipetting, user error. These proven features are standard on all xTIP® Low Retention Pipette Tips, maximizing accuracy and precision.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty