Would you like to log in again?
xTIP® TipEject™ Pipette Tip Reload System for Rainin LTS Style Pipettes, Biotix
xTIP®
R-1000-9RC R-0020-9RC R-0250-9RC
10834-106CS 196.16 USD InStock
10834-106 10834-104 10834-088
xTIP® TipEject™ Pipette Tip Reload System for Rainin LTS Style Pipettes, Biotix
High quality, low retention tips are manufactured under a stringent quality control system. These tips help to reduce insertion forces, help the environment, and help improve laboratory accuracy and precision.
  • Clear casing and colored snap cards for easy tip identification
  • Operated easily with one hand
  • Helps to reduce waste by 80%
  • Tips are shielded from the environment and user handling
  • Contains all the advantages of Biotix® Tips, including FlexFit®, X-Resin®, and Blade® technologies
TipEject™ Reload System offers convenience and eco-friendliness for any lab
xTIP® Pipette Tips feature innovations beyond ergonomics. FlexFit®, X-Resin®, and Blade® technologies developed by Biotix are found exclusively in Biotix and xTIP® brand pipette tips. These technologies minimize inaccuracies associated with the number one issue in pipetting: user error. These proven features are standard on all xTIP® Low Retention Pipette Tips, maximizing accuracy and precision.
Tips are certified free of RNase, DNase, Pyrogens, and PCR Inhibitors. Manufactured under stringent controls in an ISO 9001 certified facility. Each and every lot is tested for precision, ensuring superior quality and compatibility. All raw materials are lot controlled.
Designed for intuitive use and engineered for simple application, TipEject™ saves time, effort, and waste. Quality sterile tips are stored in translucent packaging, fully protected from exposure to environment and handling. Made from sturdy polymer construction, TipEject™ is 100% recyclable. Included in each TipEject™ package are 10 trays (96 tips/tray) and a rack base. Part number and lot number are printed on the rack base for easy identification and traceability.
Certifications: ISO 9001.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty