Synonyms:
1-(2,5-Dioxoimidazolidin-4-yl)urea
A0211-500G
A0211-25G
TCA0211-025GEA
35.5
USD
InStock
TCA0211-025G
TCA0211-500G
Allantoin ≥98.0% (by titrimetric analysis)
Allantoin
CAS Number: 97-59-6
MDL Number: MFCD00005260
Molecular Formula: C4H6N4O3
Molecular Weight: 158.12
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 223
Product Categories: Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Others (Phytochemicals)
Other Product Names / Synonyms: 5-Ureidohydantoin