Synonyms:
Acetol Acetate
, Acetoxyacetone
A1066-25ML
TCA1066-025MLEA
131.95
USD
InStock
TCA1066-025ML
Acetoxy-2-propanone ≥97.0% stabilized
Acetoxy-2-propanone
(stabilized with Mg6Al2(OH)16CO3.4H2O)
CAS Number: 592-20-1
MDL Number: MFCD00042848
Molecular Formula: C5H8O3
Molecular Weight: 116.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 71
Specific Gravity (20/20): 1.08
Other Product Names / Synonyms: Acetol Acetate, Acetoxyacetone