A1406-25G
TCA1406-025GEA
82.22
USD
InStock
TCA1406-025G
5-Amino-2,4,6-triiodoisophthalic acid ≥96.0% (by titrimetric analysis)
5-Amino-2,4,6-triiodoisophthalic acid
CAS Number: 35453-19-1
MDL Number: MFCD00190167
Molecular Formula: C8H4I3NO4
Molecular Weight: 558.84
Purity/Analysis Method: 96.0% (T)
Form: Crystal
Melting point (°C): 270