Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) ≥98.0% (by HPLC)

Supplier: TCI America
Synonyms: 3H-1,2-Benzodithiol-3-one-1,1-dioxide3H-1,2-Benzodithiol-3-one 1,1-Dioxide

B3125-1G B3125-5G
TCB3125-1GEA 84.25 USD InStock
TCB3125-1G TCB3125-5G
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) ≥98.0% (by HPLC)
3H-1,2-Benzodithiol-one 1,1-dioxide (Beaucage Reagent)
CAS Number: 66304-01-6
MDL Number: MFCD00132960
Molecular Formula: C7H4O3S2
Molecular Weight: 200.23
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 104

Product Categories: Synthetic Organic Chemistry, Others (Sulfur Compounds for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: Beaucage Reagent


https://us-legacy.vwr.com/stibo/web/std.lang.all/45/63/28934563.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty