Synonyms:
4-Chlorobenzohydrazide
, 4-Chlorobenzoic hydrazide
, 4-Chlorobenzoic acid hydrazide
, 4-Chlorobenzene-1-carbohydrazide
C2060-5G
C2060-25G
TCC2060-25GEA
204.02
USD
InStock
TCC2060-25G
TCC2060-5G
4-Chlorobenzhydrazide ≥98.0% (by GC, titration analysis)
4-Chlorobenzhydrazide
CAS Number: 536-40-3
MDL Number: MFCD00007603
Molecular Formula: C7H7ClN2O
Molecular Weight: 170.60
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 166
Other Product Names / Synonyms: 4-Chlorobenzoylhydrazine, 4-Chlorobenzoic Hydrazide