D1902-10G
D1902-25G
TCD1902-025GEA
328
USD
InStock
TCD1902-025G
TCD1902-010G
4,4'-Diacetylbiphenyl ≥99.0%
4,4'-Diacetylbiphenyl
CAS Number: 787-69-9
MDL Number: MFCD00017248
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: 99.0% (GC)
Form: Crystal
Melting point (°C): 194
Product Categories: Materials Chemistry, Biphenyls (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4',4''-Biacetophenone