Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichloro-4-pyridone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 3,5-Dichloropyridin-4(1H)-one

D2103-25G
TCD2103-025GEA 148.27 USD InStock
TCD2103-025G
3,5-Dichloro-4-pyridone ≥98.0% (by titrimetric analysis)
3,5-Dichloro-4-pyridone
CAS Number: 17228-70-5
MDL Number: MFCD00051673
Molecular Formula: C5H3Cl2NO
Molecular Weight: 163.99
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 325

Other Product Names / Synonyms: 3,5-Dichloro-4-pyridinol, 3,5-Dichloro-4(1H)-pyridinone, 3,5-Dichloro-4-pyridone


https://us-legacy.vwr.com/stibo/web/std.lang.all/88/84/28978884.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty