Synonyms:
Methyl 4,5-dimethoxyanthranilate
, 2-Amino-4,5-dimethoxybenzoic acid methyl ester
, Methyl 2-amino-4,5-dimethoxybenzoate
, 2-Amino-4,5-dimethoxy benzoate
D2749-25G
TCD2749-25GEA
54
USD
InStock
TCD2749-25G
Methyl-2-amino-4,5-dimethoxybenzoate ≥98.0% (by GC, titration analysis)
Methyl 2-amino-4,5-dimethoxybenzoate
CAS Number: 26759-46-6
MDL Number: MFCD00014904
Molecular Formula: C10H13NO4
Molecular Weight: 211.22
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 129
Other Product Names / Synonyms: 2-Amino-4,5-dimethoxybenzoic Acid Methyl Ester, 6-Aminoveratric Acid Methyl Ester, 4,5-Dimethoxyanthranilic Acid Methyl Ester, Methyl 2-Amino-4,5-dimethoxybenzoate, Methyl 6-Aminoveratrate