Synonyms:
N'-Acetylacetohydrazide
D2796-25G
TCD2796-25GEA
92.01
USD
InStock
TCD2796-25G
N,N'-Diacetylhydrazine ≥98.0%
N,N'-Diacetylhydrazine
CAS Number: 3148-73-0
MDL Number: MFCD00008673
Molecular Formula: C4H8N2O2
Molecular Weight: 116.12
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 139