Synonyms:
2,4-Difluorophenylisocyanate
D3086-5G
TCD3086-5GEA
114.68
USD
InStock
TCD3086-5G
2,4-Difluorophenyl isocyanate ≥98.0%
2,4-Difluorophenyl isocyanate
CAS Number: 59025-55-7
MDL Number: MFCD00001997
Molecular Formula: C7H3F2NO
Molecular Weight: 155.10
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 50
Flash Point (°C): 55
Specific Gravity (20/20): 1.33
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 2,4-Difluorophenyl Ester