Synonyms:
1,4-Diphenylbutane-1,4-dione
, 1,4-Diphenyl-1,4-butanedione
D3179-1G
TCD3179-1GEA
55.73
USD
InStock
TCD3179-1G
1,2-Dibenzoylethane ≥98.0%
1,2-Dibenzoylethane
CAS Number: 495-71-6
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 148
Other Product Names / Synonyms: 1,2-Dibenzoylethane