Synonyms:
Dimethyldiselenide
D3290-1G
D3290-5G
TCD3290-1GEA
56.82
USD
InStock
TCD3290-1G
TCD3290-5G
Dimethyl diselenide ≥98.0%
Dimethyl diselenide
CAS Number: 7101-31-7
Molecular Formula: C2H6Se2
Molecular Weight: 187.99
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 155
Melting point (°C): -82
Flash Point (°C): 60
Specific Gravity (20/20): 1.99
Product Categories: Synthetic Organic Chemistry, Selenenylation
Other Product Names / Synonyms: Methyl Diselenide