Synonyms:
Manganese(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate
, Manganese Disodium Ethylenediaminetetraacetate
E0095-25G
TCE0095-025GEA
89.29
USD
InStock
TCE0095-025G
Ethylenediaminetetraacetic acid manganese disodium salt hydrate ≥95.0% (by titrimetric analysis)
Ethylenediaminetetraacetic acid manganese disodium salt hydrate
CAS Number: 15375-84-5
MDL Number: MFCD00054451
Molecular Formula: C10H16N2O8
Molecular Weight: 389.13
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Other Product Names / Synonyms: Manganese Disodium Ethylenediaminetetraacetate