Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2,4-Pentanedionato)lithium ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: Lithium AcetylacetonateAcetylacetone Lithium SaltLithium acetylacetonate

L0080-25G L0080-500G
TCL0080-500GEA 360.06 USD InStock
TCL0080-500G TCL0080-025G
(2,4-Pentanedionato)lithium ≥98.0% (by titrimetric analysis)
(2,4-Pentanedionato)lithium
CAS Number: 18115-70-3
MDL Number: MFCD00013496
Molecular Formula: C5H7LiO2
Molecular Weight: 106.05
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 250

Other Product Names / Synonyms: Acetylacetone Lithium Salt, Lithium Acetylacetonate


Formula: C₅H₇LiO₂
MW: 106.05 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00013496
CAS Number: 18115-70-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/30/73/28973073.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty