M0774-1G
M0774-5G
TCM0774-001GEA
93.35
USD
InStock
TCM0774-001G
TCM0774-5G
4-Methyl-1H-pyrazole ≥98.0%
4-Methyl-1H-pyrazole
CAS Number: 7554-65-6
MDL Number: MFCD00005245
Molecular Formula: C4H6N2
Molecular Weight: 82.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 207
Flash Point (°C): 96
Freezing point (°C): 25