Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3',4'-(Methylenedioxy)acetophenone ≥98.0%

Supplier: TCI America

M0874-25G
TCM0874-025GEA 145.15 USD InStock
TCM0874-025G
3',4'-(Methylenedioxy)acetophenone ≥98.0%
3',4'-(Methylenedioxy)acetophenone
CAS Number: 3162-29-6
MDL Number: MFCD00005831
Molecular Formula: C9H8O3
Molecular Weight: 164.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 159
Melting point (°C): 88
Flash Point (°C): 110

Other Product Names / Synonyms: 5-Acetyl-1,3-benzodioxole


https://us-legacy.vwr.com/stibo/web/std.lang.all/43/81/28974381.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty