Synonyms:
ANPO
N0068-1G
TCN0068-001GEA
58.82
USD
InStock
TCN0068-001G
2-(1-Naphthyl)-5-phenyloxazole ≥99.0% (by HPLC, total nitrogen)
2-(1-Naphthyl)-5-phenyloxazole
[for scintillation spectrometry]
CAS Number: 846-63-9
MDL Number: MFCD00019714
Molecular Formula: C19H13NO
Molecular Weight: 271.32
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 106
Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: ANPO