Synonyms:
2-((5-Nitrofuran-2-yl)methylene)hydrazinecarboxamide
, 5-Nitrofurfural Semicarbazone
, 5-Nitro-2-Furaldehyde Semicarbazone
N0200-250G
N0200-25G
TCN0200-025GEA
19.29
USD
InStock
TCN0200-025G
TCN0200-250G
Nitrofurazone ≥98.0% (by HPLC and UV method)
Nitrofurazone
CAS Number: 59-87-0
MDL Number: MFCD00003225
Molecular Formula: C6H6N4O4
Molecular Weight: 198.14
Purity/Analysis Method: 98.0% (HPLC,E)
Form: Crystal
Melting point (°C): 244
Other Product Names / Synonyms: 5-Nitrofurfural Semicarbazone