Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Norvaline ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: H-Nva-OHL(+)-Norvaline(S)-2-Aminopentanoic acid(S)-(+)-Norvaline

N0686-5G N0686-1G
TCN0686-5GEA 115.23 USD InStock
TCN0686-5G TCN0686-1G
L-Norvaline ≥99.0% (by titrimetric analysis)
L-Norvaline
CAS Number: 6600-40-4
MDL Number: MFCD00064421
Molecular Formula: C5H11NO2
Molecular Weight: 117.15
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Specific rotation [a]20/D: 25 deg (C=10, 6mol/L HCl)

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: (S)-2-Aminopentanoic Acid, (S)-2-Aminovaleric Acid


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00064421
CAS Number: 6600-40-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/22/28989922.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty