Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H-Nonadecafluoro-1-decanol ≥98.0%

Supplier: TCI America
Synonyms: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluor-1-decanol1H,1H-Perfluoro-1-decanol2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluoro-1-decanol

N0814-5G
TCN0814-5GEA 228 USD InStock
TCN0814-5G
1H,1H-Nonadecafluoro-1-decanol ≥98.0%
1H,1H-Nonadecafluoro-1-decanol
CAS Number: 307-37-9
MDL Number: MFCD00153234
Molecular Formula: C10H3F19O
Molecular Weight: 500.10
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 136
Melting point (°C): 88

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H-Perfluoro-1-decanol


Formula: C₁₀H₃F₁₉O
MW: 500.1 g/mol
Boiling Pt: 136 °C
Melting Pt: 88 °C
MDL Number: MFCD00153234
CAS Number: 307-37-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/00/12/28990012.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty