Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyvinylpyrrolidone, insoluble

Supplier: TCI America
Synonyms: PVPPolyvidone Polyvinylpyrrolidon

P0691-100G
TCP0691-100GEA 86.89 USD InStock
TCP0691-100G
Polyvinylpyrrolidone, insoluble
Polyvinylpyrrolidone
CAS Number: 9003-39-8
Form: Crystals
Color: White

Product Categories: Materials Chemistry, Polymer Research Reagents


Formula: (C₆H₉NO)n
MDL Number: MFCD01076626
CAS Number: 9003-39-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty