Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenol red 0.1% in ethanol (20%)

Supplier: TCI America
Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

P0703-500ML
TCP0703-500MLEA 62.93 USD InStock
TCP0703-500ML
Phenol red 0.1% in ethanol (20%)
Phenol red
[for Titration]
CAS Number: 143-74-8
MDL Number: MFCD00003552
Molecular Weight: 354.38
Form: Clear Liquid
Color: Yellow
Flash Point (°C): 36


Formula: C₁₉H₁₄O₅S
MW: 354.38 g/mol
Flash Pt: 36 °C
MDL Number: MFCD00003552
CAS Number: 143-74-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/07/28991407.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty