Synonyms:
Sodium propane-1,3-disulphonate
, Sodium propane-1,3-disulfonate
P1205-5G
TCP1205-005GEA
35.49
USD
InStock
TCP1205-005G
1,3-Propanedisulfonic acid disodium salt ≥97.0% (by titrimetric analysis)
1,3-Propanedisulphonic acid disodium salt
CAS Number: 36589-58-9
MDL Number: MFCD00191483
Molecular Formula: C3H8O6S2
Molecular Weight: 248.17
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Color: White
Other Product Names / Synonyms: 1,3-Propanedisulfonic Acid Disodium Salt