Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Phenoxybenzonitrile ≥98.0%

Supplier: TCI America

P1649-5G
TCP1649-5GEA 97.44 USD InStock
TCP1649-5G
4-Phenoxybenzonitrile ≥98.0%
4-Phenoxybenzonitrile
CAS Number: 3096-81-9
Molecular Formula: C13H9NO
Molecular Weight: 195.22
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 174
Flash Point (°C): 110

Other Product Names / Synonyms: 4-Cyanodiphenyl Ether


Formula: C₁₃H₉NO
MW: 195.22 g/mol
Boiling Pt: 174 °C
Flash Pt: 110 °C
MDL Number: MFCD00017346
CAS Number: 3096-81-9
UN: 3439
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/21/92/28992192.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty