Synonyms:
1,10-Phenanthroline hydrate
P1826-5G
TCP1826-5GEA
47.71
USD
InStock
TCP1826-5G
1,10-Phenanthroline monohydrate ≥99.0% (by titrimetric analysis) for biochemical research
1,10-Phenanthroline monohydrate
[for Biochemical Research]
CAS Number: 5144-89-8
MDL Number: MFCD00149973
Molecular Formula: C12H8N2
Molecular Weight: 180.21
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Color: White
Melting point (°C): 102