Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)ruthenium

Supplier: TCI America
Synonyms: Bis(cyclopentadienyl)ruthenium(II)Ruthenocene

R0078-1G
TCR0078-1GEA 181.28 USD InStock
TCR0078-1G
Bis(cyclopentadienyl)ruthenium
Bis(cyclopentadienyl)ruthenium
CAS Number: 1287-13-4
MDL Number: MFCD00001442
Molecular Formula: C10H10Ru
Molecular Weight: 231.26
Form: Crystal
Color: Slightly Pale Yellow
Melting point (°C): 202

Other Product Names / Synonyms: Bis(cyclopentadienyl)ruthenium(II)


Formula: C₁₀Ru
MW: 221.18 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00001442
CAS Number: 1287-13-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/37/82/28993782.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty