Synonyms:
Sulfadimidine
, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulfonamide
, Sulphadimidine
, 4,6-Dimethylsulfadiazine
, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulphonamide
, 4,6-Dimethylsulphadiazine
S0586-250G
S0586-25G
TCS0586-250GEA
361.34
USD
InStock
TCS0586-250G
TCS0586-25G
Sulfamethazine ≥98.0% (by HPLC, titration analysis)
Sulfamethazine
CAS Number: 57-68-1
MDL Number: MFCD00006066
Molecular Formula: C12H14N4O2S
Molecular Weight: 278.33
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Color: White
Melting point (°C): 199
Storage Temperature: 0-10°C
Product Categories: Bioscience, Synthetic Antimicrobial Ingredients
Other Product Names / Synonyms: Sulfadimidine, 4-Amino-N-(4,6-dimethyl-2-pyrimidyl)benzenesulfonamide, 4,6-Dimethylsulfadiazine